Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer), 1 KIT
Catalog # B52
Supplier: Thermo Fisher Scientific
Buffer, TBE solution 10X concentrate (TRIS-borate-EDTA buffer)
Catalog # B52
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pakn:1 KIT
- Beskrivelse:TBE buffer solution 10X concentrate (TRIS-borate-EDTA buffer)