Tannin
Glass bottle for solids, 100 g
Katalog # SERA35753.01
Marketsource
Leverandør: Serva
Gå til:
Product Details & Documents
Synonymer: Gallotannin, Tannin
- CAS nummer:1401-55-4
- Formel:C₇₆H₅₂O₄₆
- MDL nummer:MFCD00066397
- Molecular Weight:1701.2 g/mol
- Merck Index:12,09221
- Smeltende Pt:218…219 °C
- Tæthed:>1 g/cm³ (20 °C)
- Flash Pt:199 °C
- EINECS:215-753-2
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g



