Colistin sulphate
Marketsource
Leverandør: Serva
Om denne vare
2 tilgængelige muligheder nedenfor
Product Details & Documents
Specifications for Product Family
Specifikationer
Specifikationer
- Catalog No:
- SERA17420.01
- SERA17420.02
- Protein/peptide name:
- Colistin sulphate
- Colistin sulphate
- CAS-nr:
- 1264-72-8
- 1264-72-8
- Opbevaring betingelser:
- Refrigerator
- Refrigerator
Specifikationer
Product Family Options
Produktinformation
- PakningPaknTilgængelighedPrice
- Specifikationer:Marketsource
More
Specifications
Cat. No.SERA17420.01Protein/peptide nameColistin sulphateCAS-nr1264-72-8Opbevaring betingelserRefrigeratorMarketsource - Specifikationer:Marketsource
More
Specifications
Cat. No.SERA17420.02Protein/peptide nameColistin sulphateCAS-nr1264-72-8Opbevaring betingelserRefrigeratorMarketsource