Gelatin
Marketsource
: Serva
- Catalog No:
- SERA22156.01
- SERA22156.02
- Protein/peptide name:
- Gelatin
- Gelatin
- Protein synonyms:
- Gelatine
- Gelatine
- CAS-nr:
- 9000-70-8
- 9000-70-8
- Opbevaring betingelser:
- Ambient
- Ambient
Product Family Options
- PakningPakn
- Marketsource
Specifications
Cat. No.SERA22156.01Protein/peptide nameGelatinProtein synonymsGelatineCAS-nr9000-70-8Opbevaring betingelserAmbientMarketsource - Marketsource
Specifications
Cat. No.SERA22156.02Protein/peptide nameGelatinProtein synonymsGelatineCAS-nr9000-70-8Opbevaring betingelserAmbientMarketsource