Gelatin
Glass bottle for solids, 500 g
SERA22151.02
Marketsource
: Serva
- Pakning:Glass bottle for solids
- Pakn:500 g
- Cat. No.:SERA22151.02
- CAS-nr:9000-70-8
- Protein/peptide name:Gelatin
- Protein synonyms:Gelatine
- Opbevaring betingelser:Ambient



