Orders LinkContactUsLinkComponent
Cellulose, Supelco®
Glass bottle for solids, 1 kg
Katalog # 22183-1KG-F
Leverandør:   Merck
Cellulose, Supelco®
Katalog # 22183-1KG-F
Leverandør:  Merck
CAS nummer:  9004-34-6
Cellulose, Supelco®
Cellulose, Supelco®
Glass bottle for solids, 1 kg
CAS nummer:  9004-34-6
Price
3.520,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Synonymer:  Microcrystallinecellulose
  • CAS nummer:9004-34-6
  • Formel:(C₁₂H₂₂O₁₁)n
  • MDL nummer:MFCD00081512
  • Molecular Weight:342.3 g/mol
  • Merck Index:13,01977
  • Smeltende Pt:260…270 °C
  • Tæthed:1,27…1.6 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:232-674-9
Specifikationer
Specifikationer
  • Pakning:
    Glass bottle for solids
  • Pakn:
    1 kg