Sample vials
Sample vials, 12,00 ml, No
Katalog # PAAR21550
Leverandør: Anton Paar
Gå til:
Product Details & Documents
These sample vials made of plastic are used with a sample changer (1/48/96 positions magazine) to automatically fill sample into the measuring instrument.
Specifikationer
- Kasket inkluderet:No
- Kapacitet:12,00 ml
- Beskrivelse:Sample vials