Orders LinkContactUsLinkComponent
Storage cabinet
Storage cabinet
Storage system, light grey, PS, 80×480×45 mm
Katalog # SEMA5187
Leverandør:   SEMADENI
Storage cabinet
Katalog # SEMA5187
Leverandør:  SEMADENI
Storage cabinet
Storage cabinet
Storage system, light grey, PS, 80×480×45 mm
Price
376,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The storage cabinet system allows a quick, individual and efficient division of drawers, shelves, cupboards, etc.
  • The boxes can be put together in a number of variations according to a sophisticated module
  • The boxes have holders on the bottom, which fit into the grid area of ​​the self-adhesive drawer foil
  • This self-adhesive foil prevents the boxes from slipping around in the drawer
  • Heavy-duty boxes made of impact-resistant polystyrene (PS).
Specifikationer
  • Beskrivelse:
    Storage system, light grey, PS
  • B×D×H:
    80×480×45 mm