Orders LinkContactUsLinkComponent
Centrifugerør, Corning®
Centrifugerør, Corning®
   Sustainable Options Available
Supplier:   Corning
Centrifugerør, Corning®
Supplier:  Corning
   Sustainable Options Available
Certificates
Centrifugerør, Corning®
7 Options Available Below
Product Details & Documents
Corning 50 ml centrifuge tubes made of PP or PET.
  • Conical, with HDPE CentriStar™ or plug seal caps or without cap, sterile
  • Black printed graduations and a large white marking spot
  • Certified non pyrogenic and DNase/RNase-free
The CentriStar™ cap with leakproof design has a easy-on/easy-off flat top and offers advanced ergonomics with its wider knurls and roll-over edge design for easier gripping.
Ordering information: Skumracks og CentriStar™-låg fås separat.
Packaging: Bulk or in racks.
Specifications for Product Family
Specifications
  • Catalog No:
  • 734-1827
  • 734-1828
  • 734-1829
  • 734-1868
  • 734-1869
  • 525-3414
  • 525-0479
  • Environmentally Preferable:
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Miljøcertificering:
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
  • I-RECs, ISO 14001 - Reynosa
Specifications
Product Family Options
With plug seal cap
Product Information
  • Kapacitet
    Materiale
    Lukkestørrelse
    RCF maks.
    Model
    Pakket
    Availability
    Price
  • 50 ml
    PP
    Skruelåg
    15500×g
    koniske
    25 stk./rack
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 1.980,00 kr
    1.980,00 kr
    1 * 500 Items
       1.980,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    734-1827
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • 50 ml
    PP
    Skruelåg
    15500×g
    koniske
    25 stk./pose
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 1.780,00 kr
    1.780,00 kr
    1 * 500 Items
       1.780,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    734-1828
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • 50 ml
    PET
    Skruelåg
    3600×g
    koniske
    25 stk./rack
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 2.860,00 kr
    2.860,00 kr
    1 * 500 Items
       2.860,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    734-1829
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
With CentriStar™ cap
Product Information
  • Kapacitet
    Materiale
    Lukkestørrelse
    RCF maks.
    Model
    Pakket
    Availability
    Price
  • 50 ml
    PP
    Skruelåg
    15500×g
    koniske
    25 stk./rack
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 1.980,00 kr
    1.980,00 kr
    1 * 500 Items
       1.980,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    734-1868
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • 50 ml
    PP
    Skruelåg
    15500×g
    koniske
    25 stk./pose
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 1.770,00 kr
    1.770,00 kr
    1 * 500 Items
       1.770,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    734-1869
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
  • 50 ml
    PP
    Skruelåg
    15500×g
    Conical
    25/rack
    Log in for availability
    Log in for availability
    1 * 300 Items
    1 * 300 Items - 1.420,00 kr
    1.420,00 kr
    1 * 300 Items
       1.420,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    525-3414
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
Uden låg
Product Information
  • Kapacitet
    Materiale
    Lukkestørrelse
    RCF maks.
    Model
    Pakket
    Availability
    Price
  • 50 ml
    PP
    uden cap
    12000×g
    koniske
    25 stk./pose
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 1.270,00 kr
    1.270,00 kr
    1 * 500 Items
       1.270,00 kr
    Specifications:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    525-0479
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    I-RECs, ISO 14001 - Reynosa
      Lav fremstillingspåvirkning
CentriStar™ caps
Product Information
  • Catalog #
    Til rør
    Materiale
    Beskrivelse
    Availability
    Price
  • 50 ml
    HDPE
    Sterile
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 934,00 kr
    934,00 kr
    1 * 500 Items
       934,00 kr
Product Information
  • Catalog #
    Til rør
    Materiale
    Beskrivelse
    Availability
    Price
  • 15 ml
    HDPE
    Sterile
    Log in for availability
    Log in for availability
    1 * 500 Items
    1 * 500 Items - 555,00 kr
    555,00 kr
    1 * 500 Items
       555,00 kr
Racks
Product Information
  • Catalog #
    Til rør
    Availability
    Price
  • 50 ml
    Log in for availability
    Log in for availability
    1 * 20 Items
    1 * 20 Items - 402,00 kr
    402,00 kr
    1 * 20 Items
       402,00 kr
Product Information
  • Catalog #
    Til rør
    Availability
    Price
  • 15+50 ml
    Log in for availability
    Log in for availability
    1 * 20 Items
    1 * 20 Items - 0,00 kr
    0,00 kr
    1 * 20 Items
       0,00 kr
Product Information
  • Catalog #
    Til rør
    Availability
    Price
  • Standard rack for 15 ml centrifuge tubes
    Log in for availability
    Log in for availability
    1 * 20 Items
    1 * 20 Items - 285,00 kr
    285,00 kr
    1 * 20 Items
       285,00 kr