Orders LinkContactUsLinkComponent
Silikoneslanger, HelixMark®
Silikoneslanger, HelixMark®
Silikoneslanger, HelixMark®
Certifikater
Silikoneslanger, HelixMark®
Silikoneslanger, HelixMark®
2 tilgængelige muligheder nedenfor
Product Details & Documents
Silicone, translucent, platinum-cured.
  • Can be sterilised by autoclaving or ethylene oxide as well as by gamma radiation within the user's validation cycles and exposure levels
  • Manufactured according to GMP in an ISO 13485 certified production facility
  • Compliant with the biocompatibility requirements of USP, Class VI
Biomedical/pharmaceutical grade tubing, manufactured by Wacker Silicones from HM-6540 platinum cured material. The special manufacturing process minimises flow degradation over extended periods.
Hardness (Shore durometer A): 60
Density: 1,16
Leveringsinformation: Tubing is double shrink-wrapped in crush resistant boxes. Material certification and lot traceability are included.
Product Family Options
Produktinformation
  • Rør int.Ø
    Vægtykkelse
    Rør udv.Ø
    Tilgængelighed
    Price
  • 4,78 mm
    — mm
    7,95 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 15 m
    1 * 15 m - 1.490,00 kr
    1.490,00 kr
    1 * 15 m
       1.490,00 kr
  • 12,70 mm
    3,18 mm
    19,05 mm
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 15 m
    1 * 15 m - 1.050,00 kr
    1.050,00 kr
    1 * 15 m
       1.050,00 kr