Flat Bags, PP
Flat bag, transparent, 0,05 mm, 400×250 mm
Katalog # 129-9173
Leverandør: KOEHLER TECHNISCHE PRODUKTEN
Gå til:
Product Details & Documents
Flat sample bag suitable for infectious materials.
Specifikationer
- Beskrivelse:Flat bag, transparent
- Dimensioner:400×250 mm
- Tykkelse:0,05 mm