General purpose thermometer
General thermometer, +10...+28 °C, 250 mm, Blue alcohol
Katalog # JUCE18310002
Leverandør: JUCHHEIM SOLINGEN
Gå til:
Product Details & Documents
General purpose thermometer used for swinging.
Specifikationer
- Beskrivelse:General thermometer
- Længde:250 mm
- Seq:01
- Påfyldning:Blue alcohol
- Område:+10...+28 °C