Browguard with visor, PP
PC visor with PP browguard, clear, PP/PC, 70×47 mm
Catalog # PRTWPW91CLR
Supplier: Portwest
Jump to:
Product Details & Documents
PC visor with PP browguard, is manufactured from impact resistant PP.
- Wheel ratchet adjustment system is designed for easy use
- Replacement visor available (PW92)
Certifications: EN 166 (1 B/3 B)
Specifications
- Beskrivelse:PC visor with PP browguard, clear
- L×B:70×47 mm
- Materiale:PP/PC