Tubes with screw cap, PP
13 ml, 101 mm, 16,5 mm
Katalog # SARS60.540.016
Marketsource
Leverandør: SARSTEDT
Gå til:
Product Details & Documents
PP, translucent, with assembled integral O-ring screw cap.
- Very good chemical resistance
- Very good temperature resistance
- Sterile
Round bottom, with attached screw cap.
Specifikationer
- Udv.Ø:16,5 mm
- Kapacitet:13 ml
- Længde:101 mm