Azotometer/saccharometer
Azotometer/saccharometer, measuring tube alone, 50 ml, 0 − 100 ml, 1/5
Katalog # NEUB0345-51-050
Leverandør: NEUBERT VOLUME GLASSWAERE
Gå til:
Product Details & Documents
Azotometer/saccharometer made of borosilicate glass, with glass base.
Specifikationer
- Beskrivelse:Azotometer/saccharometer, measuring tube alone, 50 ml
- Seq:04
- Inddeling:1/5
- Område:0 − 100 ml