Microfiltration assembly, ULTRA-WARE®, KIMBLE®
Microfiltration assembly with GL 45 thread
Katalog # SCERSP179950-4532
Leverandør: DWK Life Sciences
Microfiltration assembly, ULTRA-WARE®, KIMBLE®
Katalog # SCERSP179950-4532
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
This microfiltration adapter assembly is designed to allow connection of a 47 or 90 mm filtration support base with a straight drip tip to a bottle or reservoir with a GL 45 thread.
Filtrate is collected directly into a media-type bottle when using a standard membrane filtration assembly.
Certifications: Glass conforms to USP Type I and ASTM E438, Type I, Class A requirements
Specifikationer
- Beskrivelse:Microfiltration assembly with GL 45 thread
- Cat. No.:SCERSP179950-4532
- Filtre:47 - 90 mm