Trio mop, SprayPro
Trio mop, hook and loop, trefarvet, 400 mm, 20 packs/case
Katalog # VILE143203
Leverandør: FHP VILEDA SCS
Gå til:
Product Details & Documents
Trio mop is ideal for heavily soiled surfaces, unstructured floors, tiles and many more. The looped yarn construction takes the shape of any floor, removing trapped dirt and the external fringes collect and remove all loose dirt particles.
- Unique triple zone construction
- White microfibre zone for high cleaning performance
- Grey zone for low friction
- Beige zone for high absorbency
- Machine washable over 250 times
It is excellent for spill pick up with the capacity to pick up over 500 ml in between each wringing process saving time.
Suitable for health care industries, general building, hotel, catering and restaurant.
Specifikationer
- Farve:trefarvet
- Beskrivelse:Trio mop, hook and loop
- Længde:400 mm
- Pakket:20 packs/case