Square cover glasses
Square cover glasses, 30×30 mm, 0,17 ±0,01 mm
Catalog # HECH92101401240
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Square cover glasses are made up of colourless borosilicate glass.
- Low levels of inclusions, striae, bubbles and streaks
- High spectral transmission, excellent flatness, optimum resistance to chemicals
- Refractive index finely adapted to microscopes
Certifications: ISO 8255
Specifications
- Beskrivelse:Square cover glasses
- L×B:30×30 mm
- Tykkelse:0,17 ±0,01 mm