VWR® RM 10, Moisture Tester
Katalog # 620-1956
Leverandør: VWR Collection
Specifikationer
- Model:RM 10
- Beskrivelse:Moisture tester
- Environmentally Preferable:
- Pakn:1
Specifikationer
Om denne vare
The RM 10 allows detection of moisture behind ceramic tiles and/or various wall or floor coverings, in wood down to a depth of 3 cm and in concrete or wash floor down to 4 cm.
- Easy and fast moisture rating, non destructive measurement
- Displayed values by „digit“ are relative so that values can be compared
- To check moisture on surfaces – No injection into the measuring object required
Suitable for high-speed determination of moisture i.e. estate agents, property management, house owners, architects, building experts, building contractors.
Caution: This is an indicator for fast estimation – Does not replace precision instruments like the RM 20.