Leather gloves, A271
Leverandør: Portwest
Om denne vare
Premium quality leather drivers glove with Insulatex™ lining for added warmth and comfort.
- Made from cow grain leather
- Provide mechanical and cold protection
Certifications: EN 388:2016 (3142X), EN 511 (22X)
2 tilgængelige muligheder nedenfor
Product Details & Documents
Premium quality leather drivers glove with Insulatex™ lining for added warmth and comfort.
- Made from cow grain leather
- Provide mechanical and cold protection
Certifications: EN 388:2016 (3142X), EN 511 (22X)
Specifications for Product Family
Specifikationer
- Catalog No:
- PRTWA271TARL
- PRTWA271TARXL
- Model:
- A271
- A271
- Ambidextrous/Hånd-specifik:
- Hånd-specifik
- Hånd-specifik
- Engangs/Genbrugelig:
- Genanvendelig
- Genanvendelig
- Foringsmateriale:
- Insulatex™
- Insulatex™
- EN norm:
- EN 388:2016 (3142X), EN 511 (22X)
- EN 388:2016 (3142X), EN 511 (22X)
Specifikationer
Product Family Options
Produktinformation
- Handske størrelseFarveTilgængelighedPrice
- Specifikationer:
More
Specifications
Cat. No.PRTWA271TARLModelA271Ambidextrous/Hånd-specifikHånd-specifikEngangs/GenbrugeligGenanvendeligForingsmaterialeInsulatex™EN normEN 388:2016 (3142X), EN 511 (22X) - Specifikationer:
More
Specifications
Cat. No.PRTWA271TARXLModelA271Ambidextrous/Hånd-specifikHånd-specifikEngangs/GenbrugeligGenanvendeligForingsmaterialeInsulatex™EN normEN 388:2016 (3142X), EN 511 (22X)