Orders LinkContactUsLinkComponent
HPLC columns, Superspher®
HPLC columns, Superspher®
Supplier:   Merck
HPLC columns, Superspher®
Supplier:  Merck
Certificates
HPLC columns, Superspher®
HPLC columns, Superspher®
27 Options Available Below
Product Details & Documents
Superspher® forbedrer separationseffektiviteten af HPLC-analyser. Denne sfæriske silicabærer med høj ydeevne, som har en gennemsnitlig partikelstørrelse på 4 μm, giver den bedste tryk/separation-ydeevne i betragtning af nutidige HPLC-systemer og ifølge teoretiske beregninger og praktisk erfaring.
Antallet af teoretiske plader for Superspher® er <100000 N/m. De er derfor altid det første valg til komplekse blandinger, som kræver stor topkapacitet.
Et bredt udvalg af modifikationer af Superspher® fås: apolære derivater (RP-8, RP-8 endcapped, RP-18, RP-18 endcapped og RP-select B) og polære derivater (Si 60).
Forkolonnerne skal tilsluttes LiChroCART® kolonner med manu-CART® kolonneholder 1,51486.0001
Product Family Options
Product Information
  • Beskrivelse
    Fase
    Partikelstørrelse
    Ø indv.
    Længde
    Availability
    Price
  • Superspher® 100 RP-18 endcapped LiChroCART®
    RP-18
    4 µm
    4,00 mm
    25 mm
    Log in for availability
    Log in for availability
    1 * 3 Items
    1 * 3 Items - 5.900,00 kr
    5.900,00 kr
    1 * 3 Items
       5.900,00 kr
  • Superspher® 100 RP-18 endcapped LiChroCART®
    RP-18
    4 µm
    4,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 6.750,00 kr
    6.750,00 kr
    1 * 1 Items
       6.750,00 kr
  • Superspher® 100 RP-18 endcapped LiChroCART®
    RP-18
    4 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 9.120,00 kr
    9.120,00 kr
    1 * 1 Items
       9.120,00 kr
  • Superspher® 100 RP-18 LiChroCART®
    RP-18
    4 µm
    3,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 8.040,00 kr
    8.040,00 kr
    1 * 1 Items
       8.040,00 kr
  • Superspher® 100 RP-18 LiChroCART®
    RP-18
    4 µm
    3,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 9.110,00 kr
    9.110,00 kr
    1 * 1 Items
       9.110,00 kr
  • Superspher® 100 RP-18 LiChroCART®
    RP-18
    4 µm
    4,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 8.400,00 kr
    8.400,00 kr
    1 * 1 Items
       8.400,00 kr
  • Superspher® 100 RP-18 LiChroCART®
    RP-18
    4 µm
    4,00 mm
    75 mm
    Log in for availability
    Log in for availability
    1 * 3 Items
    1 * 3 Items - 9.230,00 kr
    9.230,00 kr
    1 * 3 Items
       9.230,00 kr
  • Superspher® 100 RP-18 LiChroCART®
    RP-18
    4 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 8.850,00 kr
    8.850,00 kr
    1 * 1 Items
       8.850,00 kr
  • Superspher® 100 RP-18 LiChroCART® (3 columns from 3 batches)
    RP-18
    4 µm
    3,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
  • Superspher® 100 RP-18 LiChroCART® (3 columns from 3 batches)
    RP-18
    4 µm
    4,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 16.980,00 kr
    16.980,00 kr
    1 * 1 Each
       16.980,00 kr
  • Superspher® 60 RP-8 endcapped LiChroCART®
    RP-8
    4 µm
    4,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 6.750,00 kr
    6.750,00 kr
    1 * 1 Items
       6.750,00 kr
  • Superspher® 60 RP-8 endcapped LiChroCART®
    RP-8
    4 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 9.120,00 kr
    9.120,00 kr
    1 * 1 Items
       9.120,00 kr
  • Superspher® 60 RP-8 LiChroCART®
    RP-8
    4 µm
    4,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 8.040,00 kr
    8.040,00 kr
    1 * 1 Items
       8.040,00 kr
  • Superspher® 60 RP-8 LiChroCART® (3 columns from 3 batches)
    RP-8
    4 µm
    4,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each
  • Superspher® 60 RP-8 LiChroCART®
    RP-8
    4 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 9.110,00 kr
    9.110,00 kr
    1 * 1 Items
       9.110,00 kr
  • Superspher® 60 RP-8 LiChroCART® (3 columns from 3 batches)
    RP-8
    4 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Each
    1 * 1 Each - 18.960,00 kr
    18.960,00 kr
    1 * 1 Each
       18.960,00 kr
  • Superspher® 60 RP-select B LiChroCART®
    RP-select B
    4 µm
    3,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 7.510,00 kr
    7.510,00 kr
    1 * 1 Items
       7.510,00 kr
  • Superspher® 60 RP-select B LiChroCART®
    RP-select B
    4 µm
    4,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 9.010,00 kr
    9.010,00 kr
    1 * 1 Items
       9.010,00 kr
  • Superspher® 60 RP-select B LiChroCART®
    RP-select B
    4 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 10.200,00 kr
    10.200,00 kr
    1 * 1 Items
       10.200,00 kr
  • Superspher® 60 RP-select B LiChroCART® (3 columns from 3 batches)
    RP-select B
    4 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items
  • Superspher® 60 RP-select B LiChroCART®
    RP-select B
    4 µm
    4,00 mm
    75 mm
    Log in for availability
    Log in for availability
    1 * 3 Items
    1 * 3 Items - 10.400,00 kr
    10.400,00 kr
    1 * 3 Items
       10.400,00 kr
  • Superspher® RP-18 endekappet LiChroCART®
    RP-18
    4 µm
    2,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 6.750,00 kr
    6.750,00 kr
    1 * 1 Items
       6.750,00 kr
  • Superspher® RP-18 endekappet LiChroCART®
    RP-18
    4 µm
    2,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 6.830,00 kr
    6.830,00 kr
    1 * 1 Items
       6.830,00 kr
  • Superspher® RP-18 endcapped LiChroCART®
    RP-18
    4 µm
    3,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 6.050,00 kr
    6.050,00 kr
    1 * 1 Items
       6.050,00 kr
  • Superspher® RP-18 endcapped LiChroCART®
    RP-18
    4 µm
    3,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 6.830,00 kr
    6.830,00 kr
    1 * 1 Items
       6.830,00 kr
  • Superspher® Si 60 LiChroCART®
    Si
    4 µm
    4,00 mm
    125 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 6.360,00 kr
    6.360,00 kr
    1 * 1 Items
       6.360,00 kr
  • Superspher® Si 60 LiChroCART®
    Si
    4 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Items
    1 * 1 Items - 7.170,00 kr
    7.170,00 kr
    1 * 1 Items
       7.170,00 kr