Thermocouple
Thermocouple for muffle furnaces, max. 1100 °C, Type K
Katalog # NABE801200000
Leverandør: NABERTHERM
Gå til:
Product Details & Documents
This thermocouple is suitable for muffle furnaces.
Specifikationer
- Beskrivelse:Thermocouple for muffle furnaces, max. 1100 °C
- Model:Type K