Orders LinkContactUsLinkComponent
Silt density index kit
Silt density index kit
Silt density index (SDI) test kit, 1
Katalog # LOVI56K001901
Leverandør:   Lovibond
Silt density index kit
Katalog # LOVI56K001901
Leverandør:  Lovibond
CAS nummer:  
Silt density index kit
Silt density index kit
Silt density index (SDI) test kit, 1
CAS nummer:  
Price
8.580,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The Silt Density Index (SDI) is a measure of the solids or particulate content of water and is of specific importance when commissioning reverse osmosis plant.
  • SDI calculations in just 15 minutes
  • Ideally suited for reverse osmosis plants
  • Enables calculation of membrane fouling time, backwash frequency and membrane lifespan
The kit utilises a membrane/flow meter system to enable SDI calculations in just 15 minutes. Sufficient membranes included for 100 tests.
Documents
Specifikationer
Specifikationer
  • Pakn:
    1
  • Beskrivelse:
    Silt density index (SDI) test kit
Specifikationer