Anti-SPHK2 Polyclonal Antibody
BSBTA01382-1
- Antistof type:Primary
- Antigen symbol:SPHK2
- Clonality:Polyclonal
- Vært:Rabbit
- Reactivity:Human,Rat,Mouse
- Størrelse:100 µg
- Pakn:100 µG
Rabbit IgG polyclonal antibody for SPHK2 detection. Tested with WB, IHC-F, ICC, FCM, Direct ELISA in Human;Mouse;Rat.
Type: Primary
Antigen: SPHK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human; Mouse; Rat