Ordreindtastning
Denmark
ContactUsLinkComponent
Anti-PECAM-1/CD31 Polyclonal Antibody
Anti-PECAM-1/CD31 Polyclonal Antibody
Catalog # BSBTPB9094
Supplier:  Boster Bio
undefined
Anti-PECAM-1/CD31 Polyclonal Antibody
Catalog # BSBTPB9094
Supplier:  Boster Bio

Specifications

  • Antistof type:
    Primary
  • Antigen symbol:
    PECAM-1/CD31
  • Clonality:
    Polyclonal
  • Vært:
    Rabbit
  • Reactivity:
    Human
  • Størrelse:
    100 µg
  • Pakn:
    100 µG

Specifications

About this item

Polyclonal antibody for CD31/PECAM1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: Flow Cytometry. Reactive species: Human. CD31/PECAM1 information: Molecular Weight: 82536 MW; Subcellular Localization: Isoform Long: Cell membrane; Single-pass type I membrane protein. Cell membrane; Lipid-anchor. Cell junction. Localizes to the lateral border recycling compartment (LBRC) and recycles from the LBRC to the junction in resting endothelial cells; Tissue Specificity: Expressed on platelets and leukocytes and is primarily concentrated at the borders between endothelial cells. Isoform Long predominates in all tissues examined. Isoform Delta12 is detected only in trachea. Isoform Delta14-15 is only detected in lung. Isoform Delta14 is detected in all tissues examined with the strongest expression in heart. Isoform Delta15 is expressed in brain, testis, ovary, cell surface of platelets, human umbilical vein endothelial cells (HUVECs), Jurkat T-cell leukemia, human erythroleukemia (HEL) and U-937 histiocytic lymphoma cell lines (at protein level).

Type: Primary
Antigen: PECAM-1/CD31
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human