Orders LinkContactUsLinkComponent
Single channel pipettes, mechanical, variable / fixed volume, Reference® 2 (General Lab Product)
Single channel pipettes, mechanical, variable / fixed volume, Reference® 2 (General Lab Product)
   Bæredygtige muligheder
Leverandør:   EPPENDORF
Single channel pipettes, mechanical, variable / fixed volume, Reference® 2 (General Lab Product)
Leverandør:  EPPENDORF
   Bæredygtige muligheder
Certifikater
Single channel pipettes, mechanical, variable / fixed volume, Reference® 2 (General Lab Product)
Single channel pipettes, mechanical, variable / fixed volume, Reference® 2 (General Lab Product)
32 tilgængelige muligheder nedenfor
Product Details & Documents
The robust Reference® 2 is the successor to the Reference® pipette and features a new design, reduced weight and unique one-button operation.
  • Single button operation and spring loaded tip cone for fast and ergonomic handling with reduced operating effort
  • Quick tip ejection with active aerosol reduction; haptic feedback prevents accidential tip ejection
  • Serial number is printed on multiple components of the pipette for heightened traceability
  • Embedded RFID chip containing all relevant data ensures complete traceability
  • Optional TrackIT software allows simple identification and documentation and provides ability to add own data (e.g. calibration time and results, location)
  • Autoclavable
Due to its reliable and accurate operation, the pipette is ideal for use with precious liquids or in any application requiring extremely high levels of accuracy.
Designed and constructed for general laboratory and research applications in Life Science. Typical applications include: Food and beverages, forensic, cosmetics, agriculture, animal studies, chemical analytics, research applications at medical and academic facilities.
Works with windows 7/8/vista.
Certifications: Without CE marking.
Ordering information: All adjustable volume pipettes come with an epT.I.P.S.® box (except the 2,5 ml, 5 ml and 10 ml versions).
Each 3-pack contains three pipettes, one set of matching eppendorf tips and one ballpoint pen.
Specifications for Product Family
Specifikationer
  • Catalog No:
  • 613-5832
  • EPPE4924000.908
  • 613-5833
  • EPPE4924000.916
  • 613-5834
  • EPPE4924000.924
  • 613-5835
  • 613-5836
  • 613-5837
  • 613-5838
  • 613-5839
  • 613-5840
  • 613-5841
  • 613-5842
  • EPPE4925000.014
  • EPPE4925000.022
  • EPPE4925000.030
  • EPPE4925000.049
  • EPPE4925000.057
  • EPPE4925000.065
  • EPPE4925000.073
  • EPPE4925000.081
  • EPPE4925000.090
  • EPPE4925000.103
  • EPPE4925000.111
  • EPPE4925000.120
  • EPPE4925000.138
  • EPPE4925000.146
  • EPPE4925000.154
  • EPPE4925000.162
  • EPPE4925000.170
  • 613-7086
  • Environmentally Preferable:
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Lav fremstillingspåvirkning
  • Miljøcertificering:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Specifikationer
Product Family Options
Variable volume
Produktinformation
  • Volumen
    Nøjagtighed
    Usikkerhed
    Farvekode
    Til pipettespidser
    Tilgængelighed
    Price
  • 0,1 - 2,5 µl
    0,1 µl: ±48 % / 0,048 µl
    0,25 µl: ±12 % / 0,03 µl
    1,25 µl: ±2,5 % / 0,031 µl
    2,5 µl: ±1,4 % / 0,035 µl
    0,1 µl: ±12 % / 0,012 µl
    0,25 µl: ±6,0 % / 0,015 µl
    1,25 µl: ±1,5 % / 0,019 µl
    2,5 µl: ±0,7 % / 0,018 µl
    mørkegrå
    epT.I.P.S.® 10 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.800,00 kr
    2.800,00 kr
    1 * 1 Items
       2.800,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-5832
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 0,5 - 10 µl
    0,5 µl: ±8,0 % / 0,04 µl
    1 µl: ±2,5 % / 0,025 µl
    5 µl: ±1,5 % / 0,075 µl
    10 µl: ±1,0 % / 0,1 µl
    0,5 µl: ±5,0 % / 0,025 µl
    1 µl: ±1,8 % / 0,018 µl
    5 µl: ±0,8 % / 0,004 µl
    10 µl: ±0,4 % / 0,04 µl
    grå
    epT.I.P.S.® 20 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.800,00 kr
    2.800,00 kr
    1 * 1 Items
       2.800,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-5833
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 2 - 20 µl
    2 µl: ±3,0 % / 0,06 µl
    10 µl: ±1,0 % / 0,10 µl
    20 µl: ±0,8 % / 0,16 µl
    2 µl: ±1,5 % / 0,03 µl
    10 µl: ±0,6 % / 0,06 µl
    20 µl: ±0,3 % / 0,06 µl
    lysegrå
    epT.I.P.S.® 20 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.800,00 kr
    2.800,00 kr
    1 * 1 Items
       2.800,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-5834
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 2 - 20 µl
    2 µl: ±5,0 % / 0,1 µl
    10 µl: ±1,2 % / 0,12 µl
    20 µl: ±1,0 % / 0,2 µl
    2 µl: ±1,5 % / 0,03 µl
    10 µl: ±0,6 % / 0,06 µl
    20 µl: ±0,3 % / 0,06 µl
    gul
    epT.I.P.S.® 200 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.800,00 kr
    2.800,00 kr
    1 * 1 Items
       2.800,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-5835
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 10 - 100 µl
    10 µl: ±3,0 % / 0,3 µl
    50 µl: ±1,0 % / 0,5 µl
    100 µl: ±0,8 % / 0,8 µl
    10 µl: ±1,0 % / 0,1 µl
    50 µl: ±0,3 % / 0,15 µl
    100 µl: ±0,2 % / 0,2 µl
    gul
    epT.I.P.S.® 200 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.800,00 kr
    2.800,00 kr
    1 * 1 Items
       2.800,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-5836
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 20 - 200 µl
    20 µl: ±2,5 % / 0,5 µl
    100 µl: ±1,0 % / 1,0 µl
    200 µl: ±0,6 % / 1,2 µl
    20 µl: ±0,7 % / 0,14 µl
    100 µl: ±0,3 % / 0,3 µl
    200 µl: ±0,2 % / 0,4 µl
    gul
    epT.I.P.S.® 200 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.800,00 kr
    2.800,00 kr
    1 * 1 Items
       2.800,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-5837
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 30 - 300 µl
    30 µl: ±2,5 % / 0,75 µl
    150 µl: ±1,0 % / 1,5 µl
    300 µl: ±0,6 % / 1,8 µl
    30 µl: ±0,7 % / 0,21 µl
    150 µl: ±0,3 % / 0,45 µl
    300 µl: ±0,2 % / 0,6 µl
    orange
    epT.I.P.S.® 300 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.800,00 kr
    2.800,00 kr
    1 * 1 Items
       2.800,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-5838
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 100 - 1000 µl
    100 µl: ±3,0 % / 3,0 µl
    500 µl: ±1,0 % / 5,0 µl
    1000 µl: ±0,6 % / 6,0 µl
    100 µl: ±0,6 % / 0,6 µl
    500 µl: ±0,2 % / 1,0 µl
    1000 µl: ±0,2 % / 2,0 µl
    blå
    epT.I.P.S.® 1000 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.800,00 kr
    2.800,00 kr
    1 * 1 Items
       2.800,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-5839
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 250 - 2500 µl
    250 µl: ±4,8 % / 12 µl
    1250 µl: ±0,8 % / 10 µl
    2500 µl: ±0,6 % / 15 µl
    250 µl: ±1,2 % / 3 µl
    1250 µl: ±0,2 % / 2,5 µl
    2500 µl: ±0,2 % / 5 µl
    rød
    epT.I.P.S.® 2500 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.800,00 kr
    2.800,00 kr
    1 * 1 Items
       2.800,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-5840
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 500 - 5000
    500 µl: ±2,4 % / 12 µl
    2500 µl: ±1,2 % / 30 µl
    5000 µl: ±0,6 % / 30 µl
    500 µl: ±0,6 % / 3 µl
    2500 µl: ±0,25 % / 6,25 µl
    5000 µl: ±0,15 % / 7,5 µl
    lilla
    epT.I.P.S.® 5000 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.800,00 kr
    2.800,00 kr
    1 * 1 Items
       2.800,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-5841
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 1000 - 10000 µl
    1000 µl: ±3,0 % / 30 µl
    5000 µl: ±0,8 % / 40 µl
    10000 µl: ±0,6 % / 60 µl
    1000 µl: ±0,6 % / 6 µl
    5000 µl: ±0,2 % / 10 µl
    10000 µl: ±0,15 % / 15 µl
    turkis
    epT.I.P.S.® 10000 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 2.800,00 kr
    2.800,00 kr
    1 * 1 Items
       2.800,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-5842
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
3-packs
Produktinformation
  • Volumen
    Nøjagtighed
    Usikkerhed
    Farvekode
    Til pipettespidser
    Tilgængelighed
    Price
  • EPPE4924000.908
    3-pack, option 1, incl. 3 Eppendorf Reference® 2 pipettes (0,5 - 10 µl, 10 - 100 µl, 100 - 1000 µl), epT.I.P.S.® boxes and ballpoint pen
    3-pack, option 1, incl. 3 Eppendorf Reference® 2 pipettes (0,5 - 10 µl, 10 - 100 µl, 100 - 1000 µl), epT.I.P.S.® boxes and ballpoint pen
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 6.560,00 kr
    6.560,00 kr
    1 * 1 Each
       6.560,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4924000.908
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • EPPE4924000.916
    3-pack, option 2, incl. 3 Eppendorf Reference® 2 pipettes (2 - 20 µl, 20 - 200 µl, 100 - 1000 µl), epT.I.P.S.® boxes and ballpoint pen
    3-pack, option 2, incl. 3 Eppendorf Reference® 2 pipettes (2 - 20 µl, 20 - 200 µl, 100 - 1000 µl), epT.I.P.S.® boxes and ballpoint pen
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 6.560,00 kr
    6.560,00 kr
    1 * 1 Each
       6.560,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4924000.916
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • EPPE4924000.924
    3-pack, option 3, incl. 3 Eppendorf Reference® 2 pipettes (100 - 1000 µl, 0,5 - 5 ml, 1 - 10 ml), epT.I.P.S.® samples and ballpoint pen
    3-pack, option 3, incl. 3 Eppendorf Reference® 2 pipettes (100 - 1000 µl, 0,5 - 5 ml, 1 - 10 ml), epT.I.P.S.® samples and ballpoint pen
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 6.560,00 kr
    6.560,00 kr
    1 * 1 Each
       6.560,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4924000.924
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
Fixed volume
Produktinformation
  • Volumen
    Nøjagtighed
    Usikkerhed
    Farvekode
    Til pipettespidser
    Tilgængelighed
    Price
  • 1 µl
    ±2,5 %
    ±1,8 %
    mørkegrå
    epT.I.P.S.® 10 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.590,00 kr
    1.590,00 kr
    1 * 1 Items
       1.590,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4925000.014
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 2 µl
    ±2,0 %
    ±1,2 %
    mørkegrå
    epT.I.P.S.® 10 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.590,00 kr
    1.590,00 kr
    1 * 1 Items
       1.590,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4925000.022
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 5 µl
    ±1,2 %
    ±0,6 %
    grå
    epT.I.P.S.® 20 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.590,00 kr
    1.590,00 kr
    1 * 1 Items
       1.590,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4925000.030
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 10 µl
    ±1,0 %
    ±0,5 %
    grå
    epT.I.P.S.® 20 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.590,00 kr
    1.590,00 kr
    1 * 1 Items
       1.590,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4925000.049
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 10 µl
    ±1,2 %
    ±0,6 %
    gul
    epT.I.P.S.® 200 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.590,00 kr
    1.590,00 kr
    1 * 1 Items
       1.590,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4925000.057
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 20 µl
    ±0,8 %
    ±0,3 %
    lysegrå
    epT.I.P.S.® 20 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.590,00 kr
    1.590,00 kr
    1 * 1 Items
       1.590,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4925000.065
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 20 µl
    ±1,0 %
    ±0,3 %
    gul
    epT.I.P.S.® 200 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.590,00 kr
    1.590,00 kr
    1 * 1 Items
       1.590,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4925000.073
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 25 µl
    ±1,0 %
    ±0,3 %
    gul
    epT.I.P.S.® 200 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.590,00 kr
    1.590,00 kr
    1 * 1 Items
       1.590,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4925000.081
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 50 µl
    ±0,7 %
    ±0,3 %
    gul
    epT.I.P.S.® 200 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.590,00 kr
    1.590,00 kr
    1 * 1 Items
       1.590,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4925000.090
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 100 µl
    ±0,6 %
    ±0,2 %
    gul
    epT.I.P.S.® 200 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.590,00 kr
    1.590,00 kr
    1 * 1 Items
       1.590,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4925000.103
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 200 µl
    ±0,6 %
    ±0,2 %
    gul
    epT.I.P.S.® 200 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.590,00 kr
    1.590,00 kr
    1 * 1 Items
       1.590,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4925000.111
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 200 µl
    ±0,6 %
    ±0,2 %
    blå
    epT.I.P.S.® 1000 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.590,00 kr
    1.590,00 kr
    1 * 1 Items
       1.590,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4925000.120
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 250 µl
    ±0,6 %
    ±0,2 %
    blå
    epT.I.P.S.® 1000 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.590,00 kr
    1.590,00 kr
    1 * 1 Items
       1.590,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4925000.138
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 500 µl
    ±0,6 %
    ±0,2 %
    blå
    epT.I.P.S.® 1000 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.590,00 kr
    1.590,00 kr
    1 * 1 Items
       1.590,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4925000.146
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 1000 µl
    ±0,6 %
    ±0,2 %
    blå
    epT.I.P.S.® 1000 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.590,00 kr
    1.590,00 kr
    1 * 1 Items
       1.590,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4925000.154
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 2000 µl
    ±0,6 %
    ±0,2 %
    rød
    epT.I.P.S.® 2500 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.590,00 kr
    1.590,00 kr
    1 * 1 Items
       1.590,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4925000.162
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
  • 2500 µl
    ±0,6 %
    ±0,2 %
    rød
    epT.I.P.S.® 2500 µl
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Items
    1 * 1 Items - 1.590,00 kr
    1.590,00 kr
    1 * 1 Items
       1.590,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    EPPE4925000.170
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
6-packs
Produktinformation
  • Volumen
    Nøjagtighed
    Usikkerhed
    Farvekode
    Til pipettespidser
    Tilgængelighed
    Price
  • 613-7086
    Eppendorf Reference® 2, 6-pack with pipette carousel 2, 1-channel, variable, including 6× epT.I.P.S.® box 2.0 and ballpoint pen, 0,1 - 2,5 µl, 0,5 - 10 µl, 2 - 20 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µl
    Eppendorf Reference® 2, 6-pack with pipette carousel 2, 1-channel, variable, including 6× epT.I.P.S.® box 2.0 and ballpoint pen, 0,1 - 2,5 µl, 0,5 - 10 µl, 2 - 20 µl, 10 - 100 µl, 20 - 200 µl, 100 - 1000 µl
    gul
    -
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 1 Each
    1 * 1 Each - 14.300,00 kr
    14.300,00 kr
    1 * 1 Each
       14.300,00 kr
    Specifikationer:
      Lav fremstillingspåvirkning
    More
    Specifications
    Cat. No.
    613-7086
    Environmentally Preferable
    Lav fremstillingspåvirkning
    Miljøcertificering
    Renewable Energy Certificate, ISO 14001
      Lav fremstillingspåvirkning
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Protection filter for 0,25 - 2,5 ml variable / 2 ml fixed volume / 2,5 ml fixed volume, colour code red, 1 set = 10 filters, 1 sleeve
    Protection filter for 0,25 - 2,5 ml variable / 2 ml fixed volume / 2,5 ml fixed volume, colour code red, 1 set = 10 filters, 1 sleeve
    EPPE4920622.009
    Log ind for tilgængelighed
    1 * 1 Items - 130,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Lower part, 1 - 10 ml variable, turquoise
    Lower part, 1 - 10 ml variable, turquoise
    EPPE4920611.007
    Log ind for tilgængelighed
    1 * 1 Items - 460,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Protection filter for 0,5 - 5 ml variable, colour code purple, 1 set = 10 filters, 1 sleeve
    Protection filter for 0,5 - 5 ml variable, colour code purple, 1 set = 10 filters, 1 sleeve
    EPPE4920623.005
    Log ind for tilgængelighed
    1 * 1 Items - 130,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Pipette holder for pipette carousel
    Pipette holder for pipette carousel
    613-2925
    Log ind for tilgængelighed
    1 * 1 Items - 140,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Protection filter for 1 - 10 ml variable, colour code turquoise, 1 set = 10 filters, 1 sleeve
    Protection filter for 1 - 10 ml variable, colour code turquoise, 1 set = 10 filters, 1 sleeve
    EPPE4920624.001
    Log ind for tilgængelighed
    1 * 1 Items - 130,00 kr