Standards for Water analysis, cyanide standard, CRM, VHG
Supplier: LGC Standards PROMOCHEM
Certificates
About this item
These solutions are intended for use as a calibration standard for cyanide (CN⁻).
Certifications: Traceable to NIST, SRM.
Frequently Bought Together
VWR®, Nitril handsker, NITRILE
Supplier: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
BD Plastipak™ Syringes
Supplier: BD Medical
Vand, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Vand, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ammoniumstandard, 1g/l NH₄ i vand ARISTAR® enkeltelement standard til ionkromatografi
Supplier: VWR Chemicals
pH combination electrodes, IntelliCAL™
Supplier: Hach
TraceCERT®, Supelco® Cyanide IC Standard
Supplier: Merck