Temperature controller, TR 28-3T
Temperature controller with cable, TR 28-3T
Katalog # 453-2108
Leverandør: GESTIGKEIT HARRY
Gå til:
Product Details & Documents
TR 28-3T is a regulating unit for high-temperature hotplates.
- Regulated by PID controller
Specifikationer
- Model:TR 28-3T
- Beskrivelse:Temperature controller with cable
- Cat. No.:453-2108
- Temperaturinterval:20...600 °C
- Vægt:2,2 kg
- Elektriske specifikationer:220..240 V; 50/60 Hz
- W×D×H:200×280×65 mm