Vacuum adapter, GL thread, PTFE, BOLA
GL 45, 22 mm
Katalog # BOHLN1656-45
Leverandør: Bohlender
Gå til:
Product Details & Documents
This vacuum adapter is made of PTFE with O-ring made of FKM for secure placement of standard filter cartridges made of glass or plastic on flasks with GL 45 thread.
- Lateral step-type solenoid made of PTFE
- Chemical resistance
- Temperature resistance from -15 to +200 °C
For vacuum filtration, the O-ring at the passageway seals the connection to the filter slide so that even a small vacuum is sufficient.
Specifikationer
- Hul Ø:22 mm
- Stik:GL 45