Orders LinkContactUsLinkComponent
Multi-element standards, common elements mix 2, VHG
Multi-element standards, common elements mix 2, VHG
Multi-element standards, common elements mix 2, VHG
This standard solution is intended for use as a certified reference material and calibration standard for inductively coupled plasma mass spectrometry (ICP-MS) or alternative techniques, such as inductively coupled plasma optical emission spectroscopy (ICP-OES), and flame or furnace atomic absorption spectroscopy (AA or GFAA).
Multi elements standards for Ag, Al, B, Ca, Co, Cr, Cu, Fe, K, Mg, Mn, Na, Ni, P, Pb, Si, Sn, Ti, V and Zn.
 : Accredited to ISO 9001, ISO 17034 and ISO/IEC 17025. Traceable to NIST and SRM.
Product Family Options
  • Beskrivelse
    Koncentration
    Matrice
    Pk Info
    Pakn
  • Common elements mix 2
    100 mg/L
    5% HCl/tr HF
    100 ml
    100 mL
    1 * 100 mL
    1 * 100 mL - 4.350,00 kr
    4.350,00 kr
    1 * 100 mL
       4.350,00 kr
  • Common elements mix 2
    100 mg/L
    5% HCl/tr HF
    500 ml
    500 mL
    1 * 500 mL
    1 * 500 mL - 8.590,00 kr
    8.590,00 kr
    1 * 500 mL
       8.590,00 kr