Orders LinkContactUsLinkComponent
VWR®, Support Rods
VWR®, Support Rods
13 mm, 914 mm
Katalog # 444-0919
Leverandør:   VWR Collection
VWR®, Support Rods
Katalog # 444-0919
Leverandør:  VWR Collection
VWR®, Support Rods
13 mm, 914 mm
Price
534,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Heavy-duty, top-of-the-line rods provide maximum strength and durability. Constructed of 303 stainless steel.
Specifikationer
  • Ø:
    13 mm
  • Længde:
    914 mm