Orders LinkContactUsLinkComponent
ATP assay kit
ATP Assay Kit, 100, 1 KIT
Katalog # ABNOKA1661
Leverandør:   Abnova
ATP assay kit
Katalog # ABNOKA1661
Leverandør:  Abnova
CAS nummer:  
ATP assay kit
ATP assay kit
ATP Assay Kit, 100, 1 KIT
CAS nummer:  
Price
5.300,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
ATP Assay Kit is for the rapid bioluminescent determination of ATP in biological samples.
 : Kit contains assay buffer (10 ml), substrate (120 µl), ATP enzyme (120 µl), ATP standard (120 µl).
Specifikationer
Specifikationer
  • Pakn:
    1 KIT
  • Beskrivelse:
    ATP Assay Kit
  • Test pr. sæt:
    100
Specifikationer