Moisture trap
Safe glass moisture trap, 32×260 mm, 0,3 kg
Katalog # PERCN9301193
Leverandør: PerkinElmer
Gå til:
Product Details & Documents
Gas contacts only glass, metal and the adsorbents for purity. The Drierite® indicator and molecular sieve 5A are packed in glass protected by an outer plastic tube in the event that the glass breaks. Unique loading design allows operation in any orientation without channeling.
Designed for GC detectors that require high purity gases and recommended for ELCD and ECD systems where moisture and contamination are possible.
Maximum pressure: 6,9 bar
Specifikationer
- Beskrivelse:Safe glass moisture trap, 32×260 mm, 0,3 kg