Orders LinkContactUsLinkComponent
Ginger, Reference Standards
Leverandør:   USP
Ginger, Reference Standards
Leverandør:  USP
Certifikater
Ginger, Reference Standards
2 tilgængelige muligheder nedenfor
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Ginger Constituent Mixture: A freeze-dried mixture of 6-Gingerol and 6-Shogaol
 : US Pharmacopea Reference Standard
Product Family Options
Produktinformation
  • Standard
    Pakning
    Pakn
    Tilgængelighed
    Price
  • 1291504.
    Powdered Ginger
    Powdered Ginger
    USP reference standard
    Vial
    500 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 500 mg
    1 * 500 mg - 4.930,00 kr
    4.930,00 kr
    1 * 500 mg
       4.930,00 kr
  • 1291446.
    Ginger Constituent Mixture
    Ginger Constituent Mixture
    USP reference standard
    Vial
    0,6 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 0,6 mg
    1 * 0,6 mg - 4.410,00 kr
    4.410,00 kr
    1 * 0,6 mg
       4.410,00 kr