Orders LinkContactUsLinkComponent
Gatifloxacin, Reference Standards
Gatifloxacin, Reference Standards
Leverandør:   USP
Gatifloxacin, Reference Standards
Leverandør:  USP
Certifikater
4 tilgængelige muligheder nedenfor
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifikationer
Specifikationer
  • Catalog No:
  • 1288408.
  • 1288419.
  • 1288441.
  • 1288452.
  • Beskrivelse:
  • Gatifloxacin
  • Gatifloxacin Related Compound A
  • Gatifloxacin Related Compound D
  • Gatifloxacin Related Compound E
  • Synonym(s):
  • 1-Cyclopropyl-6-fluoro-8-hydroxy-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
  • 1-Cyclopropyl-6-fluoro-8-methoxy-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid
  • 1-Cyclopropyl-6-fluoro-8-methoxy-7-(2-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid monohydrochloride
  • CAS-nr:
  • 112811-59-3
  • 616205-76-6
  • 112811-57-1
  • 1219097-41-2
  • Formel:
  • C₃₈H₅₀F₂N₆O₁₁
Specifikationer
Product Family Options
Produktinformation
  • Standard
    Pakning
    Pakn
    Tilgængelighed
    Price
  • 1288408.
    Gatifloxacin
    Gatifloxacin
    USP reference standard
    Vial
    300 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 300 mg
    1 * 300 mg - 3.350,00 kr
    3.350,00 kr
    1 * 300 mg
       3.350,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1288408.
    Beskrivelse
    Gatifloxacin
    Synonym(s)
    CAS-nr
    112811-59-3
    Formel
    C₃₈H₅₀F₂N₆O₁₁
  • 1288419.
    Gatifloxacin Related Compound A
    Gatifloxacin Related Compound A
    USP reference standard
    Vial
    15 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 15 mg
    1 * 15 mg - 8.520,00 kr
    8.520,00 kr
    1 * 15 mg
       8.520,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1288419.
    Beskrivelse
    Gatifloxacin Related Compound A
    Synonym(s)
    1-Cyclopropyl-6-fluoro-8-hydroxy-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
    CAS-nr
    616205-76-6
    Formel
  • 1288441.
    Gatifloxacin Related Compound D
    Gatifloxacin Related Compound D
    USP reference standard
    Vial
    15 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 15 mg
    1 * 15 mg - 9.030,00 kr
    9.030,00 kr
    1 * 15 mg
       9.030,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1288441.
    Beskrivelse
    Gatifloxacin Related Compound D
    Synonym(s)
    1-Cyclopropyl-6-fluoro-8-methoxy-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid
    CAS-nr
    112811-57-1
    Formel
  • 1288452.
    Gatifloxacin Related Compound E
    Gatifloxacin Related Compound E
    USP reference standard
    Vial
    15 mg
    Log ind for tilgængelighed
    Log ind for tilgængelighed
    1 * 15 mg
    1 * 15 mg - 9.680,00 kr
    9.680,00 kr
    1 * 15 mg
       9.680,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1288452.
    Beskrivelse
    Gatifloxacin Related Compound E
    Synonym(s)
    1-Cyclopropyl-6-fluoro-8-methoxy-7-(2-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid monohydrochloride
    CAS-nr
    1219097-41-2
    Formel
Recommendations