Lockout Satchel
Lockout satchel, nylon, rød, 380×100×355 mm
Catalog # BRDY806200
Supplier: Brady
Jump to:
Product Details & Documents
Lockout satchel is made from 1000 denier red cordura nylon with a 95 cm shoulder strap.
Delivery information: Includes latchable front pocket and two side mesh holders.
Specifications
- Farve:rød
- Beskrivelse:Lockout satchel, nylon
- B×D×H:380×100×355 mm