Orders LinkContactUsLinkComponent
Single pole circuit breaker lockouts
 :   Brady
Single pole circuit breaker lockouts
 :  Brady
Single pole circuit breaker lockouts
Single pole circuit breaker lockouts
Designed for 120 V breakers that have holes in the switch tongue.
  • Quick and easy - just snap in place and apply lock
  • Dual cavities accommodate different size switch tongues
  • One padlock can be applied
Max. shackle diameter: 7 mm
Min. shackle clearance: 20 mm
Product Family Options
  • Model
    Farve
    B×D×H
  • Single pole circuit breaker lockout
    rød
    54,5×8,4×21 mm
    1 * 1 Items
    1 * 1 Items - 122,00 kr
    122,00 kr
    1 * 1 Items
       122,00 kr
  • Single pole circuit breaker lockout
    rød
    54,5×8,4×15,2 mm
    1 * 6 Items
    1 * 6 Items - 658,00 kr
    658,00 kr
    1 * 6 Items
       658,00 kr