Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human Recombinant GMFB (from <i>E. coli</i>)

Human Recombinant GMFB (from E. coli)

50 µG
Katalog # PRSI92-292
Leverandør:   ProSci Inc.
Human Recombinant GMFB (from <i>E. coli</i>)
Katalog # PRSI92-292
Leverandør:  ProSci Inc.
CAS nummer:  
Human Recombinant GMFB (from E. coli)

50 µG

CAS nummer:  
Price
2.150,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Glia maturation factor beta (GMFB) contains a ADF-H domain,which is a member of the actin-binding proteins ADF family, GMF subfamily. It is a nerve growth factor implicated in nervous system development, angiogenesis and immune function. GMFB causes differentiation of brain cells, stimulation of neural regeneration, and inhibition of proliferation of tumor cells. It is phosphorylated after phorbol ester stimulation, and is crucial for the nervous system. GMFB overexpression in astrocytes results in the increase of BDNF production. GMFB expression is increased by exercise, thus BDNF is important for exercise-induction of BDNF.

Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Documents

Specifikationer

Specifikationer

  • Pakn:
    50 µG
  • Protein/peptide name:
    GMFB
  • Protein synonyms:
    GMF-beta|Glia maturation factor beta|GMFB
  • Cat. No.:
    PRSI92-292
  • Protein/peptide type:
    Recombinant
  • Sekvens:
    Met1-His142
  • Molekylvægt:
    17.7 kD
  • Formulering:
    Lyophilized from a 0.2 um filtered solution of 20mM Tris,200mMNaCl,pH8.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tag sequence:
    C-6 His tag
  • Renhed:
    > 95% as determined by reducing SDS-PAGE
  • Tested applications:
    Bioassay
  • Kilde:
    E. coli
  • Species:
    Human
  • Opbevaring betingelser:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.