Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human recombinant CLEC3B (from cells)

Human recombinant CLEC3B (from cells)

50 µG
Katalog # PRSI91-409
Leverandør:   ProSci Inc.
Human recombinant CLEC3B (from cells)
Katalog # PRSI91-409
Leverandør:  ProSci Inc.
CAS nummer:  
Human recombinant CLEC3B (from cells)

50 µG

CAS nummer:  
Price
3.370,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

C-Type Lectin Domain Family 3 Member B (CLEC3B) is a serum and tissue protein and it contais a C-type lectin which binds to Ca++. CLEC3B is originally found in plasma, the concentrations approximately 10mg/l. It can bind to kringle 4 of plasminogen and enhance the activation of plaminogen to plasmin, catalyzed by tissue plasminogen activator in the presence of poly-D-lysine. In addition, CLEC3B may be involved in the packaging of molecules destined for exocytosis. Also, CLEC3B is best known as a prognostic marker in ovarian cancer.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

Documents

Specifikationer

Specifikationer

  • Pakn:
    50 µG
  • Protein/peptide name:
    CLEC3B
  • Protein synonyms:
    Plasminogen Kringle 4-Binding Protein|TNA|C-Type Lectin Domain Family 3 Member B|TN|Tetranectin|CLEC3B
  • Cat. No.:
    PRSI91-409
  • Protein/peptide type:
    Recombinant
  • Sekvens:
    Glu22-Val202
  • Molekylvægt:
    21.21 kD
  • Formulering:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tag sequence:
    C-6 His tag
  • Renhed:
    > 95% as determined by reducing SDS-PAGE
  • Tested applications:
    For Biological Assays
  • Kilde:
    Cells
  • Species:
    Human
  • Opbevaring betingelser:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.