Anti-TTC19 Rabbit Polyclonal Antibody
100 µg
Katalog # PRSI29-197
Leverandør: ProSci Inc.
Gå til:
Product Details & Documents
cDNA library was prepared and sequenced in Mouse Genome Encyclopedia Project of Genome Exploration Research Group in Riken Genomic Sciences Center and Genome Science Laboratory in RIKEN.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: TTC19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Documents
10107-748.pdf
Datablad
Specifikationer
- Størrelse:100 µg
- Western blot:Yes
- Cat. No.:PRSI29-197
- Clonality:Polyclonal
- Reactivity:Rat|Mouse
- Konjugation:Unconjugated
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of mouse TTC19.
- Vært:Rabbit
- Antigen symbol:TTC19
- Antigen name:Tetratricopeptide Repeat Doman 19
- ELISA:Yes



