Elemental analysis standards, flint clay - constituent elements, NIST-97B
NIST-97B, Flint clay - constituent elements, 60 g
Katalog # PROONIST-97B
Leverandør: LGC Standards PROMOCHEM
Elemental analysis standards, flint clay - constituent elements, NIST-97B
Katalog # PROONIST-97B
Leverandør: LGC Standards PROMOCHEM
CAS nummer:
Gå til:
Product Details & Documents
This standard reference material is intended for use in the determination of constituent elements in clay and material of similar matrix. Powdered clay was air-dried, ball-milled and blended to ensure homogeneity.
Specifikationer
Specifikationer
- Pakn:60 g
- Beskrivelse:NIST-97B, Flint clay - constituent elements