BugBuster® Ni-NTA His•Bind® purification kit
Catalog # 70793-3
Supplier: Merck Millipore (Novagen)
CAS Number:
Specifications
- Pakn:1 KIT
- Beskrivelse:BugBuster® HisBind® Purification Kit
Specifications
About this item
BugBuster®Ni-NTA HIS-BIND® Purification Kit is used for protein purification. Ni-NTA HIS-BIND® Resin is high-performance Ni2+-charged agarose used for rapid, one-step purification of proteins containing a polyhistidine tag sequence.
BugBuster®Ni-NTA HIS-BIND® Purification Kit has been used for the purification of His tagged proteins such as trehalose-6-phosphate phosphatase (TPP) , Ras(WT) proteins and CsgA (major curlin subunit) proteins.
Composants
2 × 100 mlBugBuster Protein Extraction Reagent
10,000 UBenzonase Nuclease, purity >90%
10 mlNi-NTA His•Bind Resin
pkg/4Chromatography Columns