Orders LinkContactUsLinkComponent
Spill control funnels, EcoPolyBlend™
Spill control funnels, EcoPolyBlend™
Funnel for nonflammables, with 9 l sump capacity
Katalog # 119-0389
Leverandør:   JUSTRITE MANUFACTURING
Spill control funnels, EcoPolyBlend™
Katalog # 119-0389
Leverandør:  JUSTRITE MANUFACTURING
Spill control funnels, EcoPolyBlend™
Funnel for nonflammables, with 9 l sump capacity
Price
894,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Spill control funnel is an eco-friendly, cost-effective way to collect liquids and keep drum tops clean.
  • Low-profile and wide diameter
  • Quick and easy pouring
  • Molded-in hook stores optional funnel cover
Scalloped, sloped design minimizes splashes and prevents containers from resting in their own liquids. Large, flat welll at the throat of funnel allows for passive draining of paint tins, filters, buckets and other containers.
OD×H: 533×83 mm; Capacity: 2 kg
Specifikationer
  • Beskrivelse:
    Funnel for nonflammables, with 9 l sump capacity