Avantor® Alltima, HPLC Columns, Cyano
Supplier: Avantor
Certificates
About this item
Avantor® Alltima HPLC columns are high quality phases designed for general purpose applications.
- Base deactivated silica phases
- Stable bonding for long column lifetime
- Multiple selectivity options
- USP L10
General purpose cyano suitable for normal or reversed phase use. Suitable for rugged normal phase applications.
Specifications
- Catalog No:
- HICH88189
- HICH88191
- HICH81189
- HICH88180
- HICH88182
- 96084/N
- Environmentally Preferable:
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Lav fremstillingspåvirkning
- Endcapped:
- Ja
- Ja
- Ja
- Ja
- Ja
- Surface area:
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- USP code:
- L10
- L10
- L10
- L10
- L10
- Miljøcertificering:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)