Wipes for delicate tasks, KIMTECH SCIENCE*
hvid, 200×205 mm, 200 tissues/box
Katalog # ROTHP502.1
Marketsource
Leverandør: Roth Carl
Gå til:
Product Details & Documents
These tissues are made of 100 % pure pulp. They are excellent for wiping smaller amounts of liquid and for cleaning pipettes and slides.
- Two-ply, white, bleached without using chlorine
- Particularly absorbent and soft
- Suitable for application with solvents and chemicals
- Low-fluffing and especially ideal for sensitive areas
Specifikationer
- Pakket:200 tissues/box
- Farve:hvid
- L×B:200×205 mm
- Seq:02