Timolol Related Compounds, Reference Standard
Om denne vare
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
6 tilgængelige muligheder nedenfor
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
Specifications for Product Family
Specifikationer
Specifikationer
- Catalog No:
- 1667381.
- 1667315.
- 1667326.
- 1667369.
- 1667443.
- 1667348.
- Beskrivelse:
- Timolol Related Compound A
- Timolol Related Compound B
- Timolol Related Compound C
- Timolol Related Compound D
- Timolol Related Compound E
- Timolol Related Compound F
- Synonym(s):
- (R)-1-(tert-Butylamino)-3-(4-morpholino-1,2,5-thiadiazol-3-yloxy)propan-2-ol maleate
- 3-(tert-Butylamino)-2-(4-morpholino-1,2,5-thiadiazol-3-yloxy)propan-1-ol
- N-(tert-Butyl)-2,3-bis(4-morpholino-1,2,5-thiadiazol-3-yloxy)propan-1-amine maleate
- 4-Morpholino-1,2,5-thiadiazol-3-ol
- (S,Z)-4-({1-(tert-butylamino)-3-[(4-morpholino-1,2,5-thiadiazol-3-yl)oxy]propan-2-yl}oxy)-4-oxobut-2-enoic acid maleate salt (1:1)
- 3-Chloro-4-morpholino-1,2,5-thiadiazol
- CAS-nr:
- 26839-77-0
- 59697-06-2
- 610271-57-3
- 30165-97-0
- 1026075-53-5
- 30165-96-9
- Formel:
- C₁₇H₂₈N₄O₇S
- C₁₃H₂₄N₄O₃S
- C₁₉H₃₁N₇O₄S₂ ·C₄H₄O₄
- C₆H₉N₃O₂S
- C₁₇H₂₆N₄O₆S
- C₆H₈ClN₃OS
Specifikationer
Product Family Options
Produktinformation
- StandardPakningPaknTilgængelighedPrice
- 1667381.Timolol Related Compound ATimolol Related Compound AUSP reference standardVial20 mgLog ind for tilgængelighed1 * 20 mg9.090,00 krSpecifikationer:
More
Specifications
Cat. No.1667381.BeskrivelseTimolol Related Compound ASynonym(s)(R)-1-(tert-Butylamino)-3-(4-morpholino-1,2,5-thiadiazol-3-yloxy)propan-2-ol maleateCAS-nr26839-77-0FormelC₁₇H₂₈N₄O₇S - 1667315.Timolol Related Compound BTimolol Related Compound BUSP reference standardVial20 mgLog ind for tilgængelighed1 * 20 mg9.540,00 krSpecifikationer:
More
Specifications
Cat. No.1667315.BeskrivelseTimolol Related Compound BSynonym(s)3-(tert-Butylamino)-2-(4-morpholino-1,2,5-thiadiazol-3-yloxy)propan-1-olCAS-nr59697-06-2FormelC₁₃H₂₄N₄O₃S - 1667326.Timolol Related Compound CTimolol Related Compound CUSP reference standardVial20 mgLog ind for tilgængelighed1 * 20 mg10.520,00 krSpecifikationer:
More
Specifications
Cat. No.1667326.BeskrivelseTimolol Related Compound CSynonym(s)N-(tert-Butyl)-2,3-bis(4-morpholino-1,2,5-thiadiazol-3-yloxy)propan-1-amine maleateCAS-nr610271-57-3FormelC₁₉H₃₁N₇O₄S₂ ·C₄H₄O₄ - 1667369.Timolol Related Compound DTimolol Related Compound DUSP reference standardVial20 mgLog ind for tilgængelighed1 * 20 mg8.850,00 krSpecifikationer:
More
Specifications
Cat. No.1667369.BeskrivelseTimolol Related Compound DSynonym(s)4-Morpholino-1,2,5-thiadiazol-3-olCAS-nr30165-97-0FormelC₆H₉N₃O₂S - 1667443.Timolol Related Compound ETimolol Related Compound EUSP reference standardVial20 mgLog ind for tilgængelighed1 * 20 mg9.500,00 krSpecifikationer:
More
Specifications
Cat. No.1667443.BeskrivelseTimolol Related Compound ESynonym(s)(S,Z)-4-({1-(tert-butylamino)-3-[(4-morpholino-1,2,5-thiadiazol-3-yl)oxy]propan-2-yl}oxy)-4-oxobut-2-enoic acid maleate salt (1:1)CAS-nr1026075-53-5FormelC₁₇H₂₆N₄O₆S - 1667348.Timolol Related Compound FTimolol Related Compound FUSP reference standardVial20 mgLog ind for tilgængelighed1 * 20 mg9.540,00 krSpecifikationer:
More
Specifications
Cat. No.1667348.BeskrivelseTimolol Related Compound FSynonym(s)3-Chloro-4-morpholino-1,2,5-thiadiazolCAS-nr30165-96-9FormelC₆H₈ClN₃OS