Orders LinkContactUsLinkComponent
Chloride FS
Chloride FS, 6×25 ml, 1 KIT
  DIAS112009990314
Chloride FS
  DIAS112009990314
 :  
Chloride FS
Chloride FS, 6×25 ml, 1 KIT
 :  
1 * 1 Each
Diagnostic reagent for quantitative in vitro determination of chloride in serum or plasma by photometric test using thiocyanate.
  • Pakn:
    1 KIT
  • Beskrivelse:
    Chloride FS, 6×25 ml