Accessories for panthera
Leverandør: MOTIC
Gå til:
Om denne vare
23 tilgængelige muligheder nedenfor
Product Details & Documents
Product Family Options
Produktinformation
- TilgængelighedPrice
- MOTI1101001100643360° rotatable analyser
- MOTI1101001101131Abbe condenser N,A, 0,90 with iris diaphragm and slot for slidersAbbe condenser N,A, 0,90 with iris diaphragm and slot for slidersLog ind for tilgængelighed1 * 1 Items685,00 kr
- MOTI1101001900631Achromatic swing-out condenser N.A, 0,90/0,13 with iris diaphragmAchromatic swing-out condenser N.A, 0,90/0,13 with iris diaphragmLog ind for tilgængelighed1 * 1 Items1.100,00 kr
- MOTI1101001901981Achromatic swing-out condenser N.A. 0,90/0,13 (strain-free) with iris diaphragmAchromatic swing-out condenser N.A. 0,90/0,13 (strain-free) with iris diaphragmLog ind for tilgængelighed1 * 1 Items1.940,00 kr
- 630-1457Darkfield slider (4×~40×)
- MOTI1101001200972Dust cover
- MOTI1101000203281EGFP/FITC/Cy2/AlexaFluor 488 set (for 630-1175/-1176)EGFP/FITC/Cy2/AlexaFluor 488 set (for 630-1175/-1176)Log ind for tilgængelighed1 * 1 Items7.700,00 kr
- MOTI1101006700192Empty insertion plate
- MOTI1101001403581Eyecup (unit)
- MOTI1101006700155Lambda tint plate
- MOTI1101011400051LED module (High color temperature, 5500 K)
- MOTI1101011400041LED module (Low color temperature, 3500 K)
- MOTI1101000300836Phase centering telescope
- MOTI1101001100391Phase slider Ph2 (20×, 40×)
- MOTI1101001901931Photo adapter (requires the photo eyepiece below)
- MOTI11010003004422,5× photo eyepiece (to be used with photo adapter)2,5× photo eyepiece (to be used with photo adapter)Log ind for tilgængelighed1 * 1 Items1.010,00 kr
- MOTI1101006701052Quartz wedge
- MOTI1101001401841Reticle with 100 divisions in 10 mm (Ø 25 mm)
- MOTI1101001401861Reticle with crossed double scale 100 divisions in 10 mm (Ø 25 mm)Reticle with crossed double scale 100 divisions in 10 mm (Ø 25 mm)Log ind for tilgængelighed1 * 1 Items379,00 kr
- MOTI1101001400141Reticle with crosshair (Ø 25 mm)
- MOTI1101001401871Reticle with pointer (Ø 25 mm)
- MOTI1101011400031Simple add on analyser
- MOTI1101006700165¹/₄ Lambda retardation plate