Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Human recombinant Renin

Human recombinant Renin

25 µG
Katalog # CAYM10006217-25
Leverandør:   Cayman Chemical
Human recombinant Renin
Katalog # CAYM10006217-25
Leverandør:  Cayman Chemical
CAS nummer:  
Human recombinant Renin

25 µG

CAS nummer:  
Price
4.650,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents

Renin is an aspartyl protease of approximately 40 kDa produced in the kidney by juxtaglomerular cells of the macula densa.

  • Renin converts angiotensinogen into angiotensin I
  • Mature and active
  • 340 amino acid protease

Native human renin is a glycoprotein derived from preprorenin by sequential cleavage of 20 and 46 amino acid residues from the N-terminus of the 406 amino acid precursor.

Documents

Specifikationer

Specifikationer

  • Pakn:
    25 µG
  • Cat. No.:
    CAYM10006217-25
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    Renin
  • Molekylvægt:
    40 kDa
  • Formulering:
    50 mM Tris, pH 8,0, containing 150 mM NaCl and 5% glycerol
  • Renhed:
    >90%
  • Species:
    Human

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.