Orders LinkContactUsLinkComponent
Tamper proof labels, safe-it
Tamper proof labels, safe-it
Tamper-proof labels, grå, 95×20 mm
Katalog # 312-0103
Leverandør:   BURKLE
Tamper proof labels, safe-it
Katalog # 312-0103
Leverandør:  BURKLE
Tamper proof labels, safe-it
Tamper proof labels, safe-it
Tamper-proof labels, grå, 95×20 mm
Price
1.230,00 kr
Log ind for tilgængelighed
1 * 1 ROL
Product Details & Documents
These safety labels are used as originality seals and as protection from manipulations of e.g. samples, retention samples, for transporting samples, sealing containers with important or dangerous chemicals in the lab as well as sealing letters, shipments and much more. The initial opening of a package is made obvious and irrevocably visible with the seal guaranteeing originality, unauthorised opening can therefore be proven.
  • With checkerboard pattern
  • Visible proof of tampering
If the safety label is removed, it divides into separate layers. A pattern that was previously hidden appears on the surface and is irreversible. The glue leaves a residue on the surface that cannot be removed (a so-called void label). Once the label has been removed, it cannot be used again. Changing the label or modifying it afterwards are therefore not possible.
When the safe-it security seal has been broken, a silver checkerboard pattern appears on both the subsurface and surface of the sticker itself.
Documents
Specifikationer
  • Farve:
    grå
  • Beskrivelse:
    Tamper-proof labels
  • Dimensioner:
    95×20 mm